Learn More
Hach Company Bottle Carrier-Linear
Supplier:  Hach Company 2094300
Encompass
Linear low-density polyethylene Securely holds up to six standard 0 5 L glass bottles pint-size reagent bottles and most other 500 mL glass or plastic bottles making breakage almost impossible Lightweight chemical-resistant and easy to clean Dimensions 34 9 x 24 8 x 27 6 cm (14 x 10 x 11in ) Maximum bottle size it will hold 73mm in diameter 178mm in height
Catalog No. 50-002-57104