Learn More
Chemglass Life Sciences Bottle, Centrifuge, 500mL, Polycarbonate, Wide Mouth
SDP
Catalog No. 501311241
Encompass_Preferred
Bottle, Centrifuge, 500mL, Polycarbonate, Wide Mouth. • Wide mouth for easy access• Useful for centrifugation, analytical testing, sampling and cell culture• Autoclavable and reusableWide-mouth, polycarbonate bottles are engineered for applications including ultracentrifugation or culture of microbes, plants, algae and organisms requiring large, surface area to volume ratios. As ultracentrifuge flasks, the bottles withstand maximum speeds without leakage and are ideal for all GS3 rotors. These centrifuge bottles are used where relatively small amounts of solids are involved. The tight fitting, polypropylene screw caps reduce the possibility of aerosol escape. Bottles and caps are reusable and resterilizable by autoclaving. The bottles are provided non sterile with caps attached as standard. The 180mL size is not graduated.
Catalog No. 50-131-1241 Supplier Chemglass Life Sciences Supplier No. CLS11560500
Bottle, Centrifuge, 500mL, Polycarbonate, Wide Mouth. • Wide mouth for easy access• Useful for centrifugation, analytical testing, sampling and cell culture• Autoclavable and reusableWide-mouth, polycarbonate bottles are engineered for applications including ultracentrifugation or culture of microbes, plants, algae and organisms requiring large, surface area to volume ratios. As ultracentrifuge flasks, the bottles withstand maximum speeds without leakage and are ideal for all GS3 rotors. These centrifuge bottles are used where relatively small amounts of solids are involved. The tight fitting, polypropylene screw caps reduce the possibility of aerosol escape. Bottles and caps are reusable and resterilizable by autoclaving. The bottles are provided non sterile with caps attached as standard. The 180mL size is not graduated.