Learn More
Foxx Life Sciences Abdos Supreme Plus Bottle Top Dispenser (10 - 100ml) 1/EA
Catalog No. NC1818683
Encompass_Preferred
Plus is the most advanced bottle top dispenser designed following years of research this next generation dispenser is highly robust and reliable. Plus has a recirculation value for redirecting the liquid into the bottle for bubble free dispensing without any loss of reagent. This Bottle Top Dispenser is made of Borosilicate Glass and has a smooth and soft plunger for easy working in difficult laboratory conditions. Solvent Resistant Borosilicate Glass Ball, Seal, and PTFE/ETP Piston.•Adjustment Knob (PP) 180º Rotation•Fully Autoclavable•Light weight and easy to use •Accuracy and Reproducibility According to ISO 8655Foxx Life Sciences is proud to represent high-quality Abdos plasticware and plastic laboratory products. For downloadable Certificates please visit: http://www.abdoslifesciences.com/Global/certificates.html.
Catalog No. NC1818683 Supplier Foxx Life Sciences Supplier No. E11716
Plus is the most advanced bottle top dispenser designed following years of research this next generation dispenser is highly robust and reliable. Plus has a recirculation value for redirecting the liquid into the bottle for bubble free dispensing without any loss of reagent. This Bottle Top Dispenser is made of Borosilicate Glass and has a smooth and soft plunger for easy working in difficult laboratory conditions. Solvent Resistant Borosilicate Glass Ball, Seal, and PTFE/ETP Piston.•Adjustment Knob (PP) 180º Rotation•Fully Autoclavable•Light weight and easy to use •Accuracy and Reproducibility According to ISO 8655Foxx Life Sciences is proud to represent high-quality Abdos plasticware and plastic laboratory products. For downloadable Certificates please visit: http://www.abdoslifesciences.com/Global/certificates.html.