Learn More
Foxx Life Sciences Abdos Supreme Duet Bottle Top Dispenser (2.5 - 30ml) 1/EA
Supplier:  Foxx Life Sciences E11734
Encompass_Preferred
Supreme Duet is the most advanced bottle top dispenser designed following years of research this next generation dispenser is highly robust and reliable. The bottle has a recirculation value for redirecting the liquid into the bottle for bubble free dispensing without any loss of reagent and rinsing the instrument without dismounting from the bottle. Duet has a smooth and soft plunger for easy working in difficult laboratory conditions. With a worldwide patent, Duet can be used in 4 modes dilution, rinsing, dual liquid handling and re-circulation all in a single unit. Solvent Resistant Borosilicate Glass Ball, Seal, and PTFE/ETP Piston. •Adjustment Knob (PP) 180º Rotation•Fully Autoclavable•Light weight and easy to use Foxx Life Sciences is proud to represent high-quality Abdos plasticware and plastic laboratory products. For downloadable Certificates please visit: http://www.abdoslifesciences.com/Global/certificates.html.
Catalog No. NC1818690