Learn More
Med Vet International Amber Plastic Dropper Bottle, 1 oz., 48/Case
Catalog No. 501180263
Encompass_Preferred
These convenient multi-purpose plastic amber bottles feature dropper caps that can be removed to administer treatment or to allow refilling. Ideal for storing liquid medications or other products that must be accurately measured and dispensed precisely with the use of included dropper pipette. Shows ounce and mL markings on bottles and mL markings on dropper. Black rubber bulb Sealing Type: Dropper cap screw closure 1 oz. (29.5 mL) capacity
Catalog No. 50-118-0263 Supplier Med Vet International Supplier No. DB1CS
These convenient multi-purpose plastic amber bottles feature dropper caps that can be removed to administer treatment or to allow refilling. Ideal for storing liquid medications or other products that must be accurately measured and dispensed precisely with the use of included dropper pipette. Shows ounce and mL markings on bottles and mL markings on dropper. Black rubber bulb Sealing Type: Dropper cap screw closure 1 oz. (29.5 mL) capacity