Learn More
Med Vet International Amber Plastic Dropper Bottle, 1 oz. 12/Box
Catalog No. 502522407
Encompass_Preferred
These convenient multi-purpose plastic amber bottles feature dropper caps that can be removed to administer treatment or to allow refilling. Ideal for storing liquid medications or other products that must be accurately measured and dispensed precisely with the use of included dropper pipette. Shows ounce and mL markings on bottles and mL markings on dropper. 12 per box. Black rubber bulb Sealing Type: Dropper cap screw closure 1 oz. (29.5 mL) capacity
Catalog No. 50-252-2407 Supplier Med Vet International Supplier No. DB1
These convenient multi-purpose plastic amber bottles feature dropper caps that can be removed to administer treatment or to allow refilling. Ideal for storing liquid medications or other products that must be accurately measured and dispensed precisely with the use of included dropper pipette. Shows ounce and mL markings on bottles and mL markings on dropper. 12 per box. Black rubber bulb Sealing Type: Dropper cap screw closure 1 oz. (29.5 mL) capacity