Learn More
Med Vet International 2 oz. Amber Plastic Dropper Bottle, 12/Box
Catalog No. 502522406
Encompass_Preferred
Convenient, multi-purpose plastic amber bottles feature dropper caps for administering treatment or refilling. Ideal for storing liquid medications or other products that must be accurately measured and dispensed precisely with the use of included dropper pipette. Shows ounce and mL markings on bottles and mL markings on dropper. 12 per box. Black rubber bulb. Sealing Type: Dropper cap screw closure. 2 oz. (59.1 mL) capacity.  
Catalog No. 50-252-2406 Supplier Med Vet International Supplier No. DB2
Convenient, multi-purpose plastic amber bottles feature dropper caps for administering treatment or refilling. Ideal for storing liquid medications or other products that must be accurately measured and dispensed precisely with the use of included dropper pipette. Shows ounce and mL markings on bottles and mL markings on dropper. 12 per box. Black rubber bulb. Sealing Type: Dropper cap screw closure. 2 oz. (59.1 mL) capacity.  