Learn More
Ace Glass, Inc. Bottle, GLS80 wide neck, 10 liter, amber, Duran 1160150, cs/1
SDP
Catalog No. 50971970
Encompass
Duran GLS80 wide neck 10L amber lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.
Catalog No. 50-971-970 Supplier Ace Glass, Inc. Supplier No. 5559220
Duran GLS80 wide neck 10L amber lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.