Learn More
Ace Glass, Inc. Bottle, GLS80 wide neck, 10 liter, clear, Duran 1113950, cs/1
SDP
Catalog No. 50971966
Encompass
Duran GLS80 wide neck 10L clear lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.
Catalog No. 50-971-966 Supplier Ace Glass, Inc. Supplier No. 555920
Duran GLS80 wide neck 10L clear lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.