Learn More
Ace Glass, Inc. Bottle, GLS80, wide neck, 10 liter, safety coated, Duran, cs/1
SDP
Catalog No. 50971974
Encompass
Duran GLS80 wide neck 10L safety coated lab bottle, IDeal for pouring, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclaveable up to 140 deg.C.
Catalog No. 50-971-974 Supplier Ace Glass, Inc. Supplier No. 5559306
Duran GLS80 wide neck 10L safety coated lab bottle, IDeal for pouring, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclaveable up to 140 deg.C.