Learn More
Ace Glass, Inc. Bottle, GLS80 wide neck, 1 liter, amber, Duran 1160147, cs/10
SDP
Catalog No. 50971969
Encompass
Duran GLS80 wide neck 1L amber lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.
Catalog No. 50-971-969 Supplier Ace Glass, Inc. Supplier No. 5559207
Duran GLS80 wide neck 1L amber lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.