Learn More
Ace Glass, Inc. Bottle, GLS80, wide neck, 20 liter, safety coated, Duran, cs/1
SDP
Catalog No. 50971975
Encompass
Duran GLS80 wide neck 20L safety coated lab bottle, IDeal for pouring, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclaveable up to 140 deg.C.
Catalog No. 50-971-975 Supplier Ace Glass, Inc. Supplier No. 5559308
Duran GLS80 wide neck 20L safety coated lab bottle, IDeal for pouring, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclaveable up to 140 deg.C.