Learn More
Ace Glass, Inc. Bottle, GLS80 wide neck, 250mL, clear, Duran 218603656, cs/10
SDP
Catalog No. 50978394
Encompass
Duran GLS80 wide neck 250mL clear lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C. 105mm x 95 mm (Height x Outer Diameter)
Catalog No. 50-978-394 Supplier Ace Glass, Inc. Supplier No. 555903
Duran GLS80 wide neck 250mL clear lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C. 105mm x 95 mm (Height x Outer Diameter)