Learn More
Ace Glass, Inc. Bottle, GLS80 wide neck, 500mL, clear, Duran 1112627, cs/10
SDP
Catalog No. 50971964
Encompass
Duran GLS80 wide neck 500mL clear lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.
Catalog No. 50-971-964 Supplier Ace Glass, Inc. Supplier No. 555905
Duran GLS80 wide neck 500mL clear lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.