Learn More
Ace Glass, Inc. Bottle, GLS80 wide neck, 5 liter, amber, Duran 1160149, cs/1
SDP
Catalog No. 50978396
Encompass
Duran GLS80 wide neck 5L amber lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.
Catalog No. 50-978-396 Supplier Ace Glass, Inc. Supplier No. 5559209
Duran GLS80 wide neck 5L amber lab bottle, IDeal for pouring, filling, mixing, etc. Allows for easy use of spoons, spatulas, tweezers, and funnels. Supplied with quick-release blue PP cap and pour ring that are autoclavable up to 140 deg.C.