Learn More
Ace Glass, Inc. Bottle, Duran, square, 1L, GL 45 blue polypropylene cap, cs/10
SDP
Catalog No. 50971978
Encompass
Duran chemically-resistant square 1L laboratory bottle, fabricated from borosilicate glass 3.3. Supplied graduated with a GL 45 blue polypropylene cap and removable pouring ring that allows drip-free operation. For use from -40 degree C up to 140 degree C. Not suitable for vacuum or pressure use. Can be hot air sterilized. Case/10.
Catalog No. 50-971-978 Supplier Ace Glass, Inc. Supplier No. 556025
Duran chemically-resistant square 1L laboratory bottle, fabricated from borosilicate glass 3.3. Supplied graduated with a GL 45 blue polypropylene cap and removable pouring ring that allows drip-free operation. For use from -40 degree C up to 140 degree C. Not suitable for vacuum or pressure use. Can be hot air sterilized. Case/10.