Learn More
Wilmad Labglass BOTTLE FILTER CARBOY 3.5 GAL
Supplier:  Wilmad Labglass LG3465102
Encompass_Preferred
Bottle, Filter Carboy Style. Capacity: 13,250mL (3- 1/2 gal. Stopper Size: no. 12; Hose Connection: 3/8 in. Large capacity bottle with hose connection sealed at neck.
Catalog No. 50-806-171