Learn More
Wilmad Labglass BOTTLE GAS WASH 1000ML COMP C
Catalog No. 50806194
Encompass_Preferred
Bottle, Gas Washing, Tall Form, with Fritted Disc. Capacity: 1000mL, Joint Size: [ST] 40/35; Porosity: Coarse. Manufactured with Inlet tube sealed through center of fritted glass disc. Inlet tube extends nearly to base of bottle.
Catalog No. 50-806-194 Supplier Wilmad Labglass Supplier No. LG3765130
Bottle, Gas Washing, Tall Form, with Fritted Disc. Capacity: 1000mL, Joint Size: [ST] 40/35; Porosity: Coarse. Manufactured with Inlet tube sealed through center of fritted glass disc. Inlet tube extends nearly to base of bottle.