Learn More
Ace Glass, Inc. Bottle, gas washing, 500mL, porosity B fritted disc, Ace-Threds
SDP
Catalog No. 50979104
Encompass_Preferred
500mL capacity Gas Washing Bottle with the convenience of Ace-Threds. The center washing tube fits into the borosilicate glass bottle via a #25 Ace-Thred with a nylon bushing and a FETFE o-ring. The washing tube has #7 Ace-Safe inlet/outlet ports with 1/4 hose connections. The bottom fritted disc is 70-100 micron porosity.
Catalog No. 50-979-104 Supplier Ace Glass, Inc. Supplier No. 716110
500mL capacity Gas Washing Bottle with the convenience of Ace-Threds. The center washing tube fits into the borosilicate glass bottle via a #25 Ace-Thred with a nylon bushing and a FETFE o-ring. The washing tube has #7 Ace-Safe inlet/outlet ports with 1/4 hose connections. The bottom fritted disc is 70-100 micron porosity.