Learn More
Chemglass Life Sciences Complete 250mL Bottle, Gas Washing, Tall Form, Coarse Frit, 305mm Approx. OAH
SDP
Catalog No. CG111414
Encompass_Preferred
Complete 250mL Bottle, Gas Washing, Tall Form, Coarse Frit, 305mm Approx. OAH. Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and coarse fritted tube for gas dispersion. Stopper is held in place with two CG-110-02, 18mm stainless steel springs. Springs are supplied when bottle is ordered complete.
Catalog No. CG111414 Supplier Chemglass Life Sciences Supplier No. CG111414
Complete 250mL Bottle, Gas Washing, Tall Form, Coarse Frit, 305mm Approx. OAH. Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and coarse fritted tube for gas dispersion. Stopper is held in place with two CG-110-02, 18mm stainless steel springs. Springs are supplied when bottle is ordered complete.