Learn More
Arizona Instrument LLC Sample Bottle, 25mL
Catalog No. NC1303223
Encompass
Glass Sample Bottle, 25ml for Vapor Pro instruments CT-3100(-L) and CT-4200XL.Â
Catalog No. NC1303223 Supplier Arizona Instrument LLC Supplier No. 9900229
Glass Sample Bottle, 25ml for Vapor Pro instruments CT-3100(-L) and CT-4200XL.Â