Learn More
BURKLE INC Bottle holder, bottles up to Ø 3.46 in. (88 mm), TeleScoop
Catalog No. NC2187707
Encompass_Preferred
Holds all bottles up to 88 mm (3.46 in.) Ø. Plastic, glass or metal bottles can be securely filled in this way. Angle steplessly adjustable up to 90°.
This item is not returnable. View return policy
Catalog No. NC2187707 Supplier BURKLE INC Supplier No. 53544100
Holds all bottles up to 88 mm (3.46 in.) Ø. Plastic, glass or metal bottles can be securely filled in this way. Angle steplessly adjustable up to 90°.