Learn More
Cole-Parmer Burkle 5354-4100 Bottle holder, PP/PA, up to 88 mm bottles
Catalog No. NC2171938 Shop All Cole-Parmer Products
Encompass_Preferred
This modular sampling system allows you to build a sampler according to your application. Select a telescoping aluminum rod (not included) with the length you need, then choose one or more beakers and/or a bottle holder to attach to the rod. Angled and pendulum beakers are available in plastic or metal, and the bottle holder fits plastic, glass, or metal bottles. Angled beakers maintain position while sampling. The polypropylene beakers feature an adjustable angle up to 90°, and the 304 SS beaker has a fixed angle with a lip—perfect for scraping caked solids, sludge, and mud. Pendulum beakers automatically swings into a horizontal position, preventing spills. This type of beaker is ideal for deep, narrow shafts and other difficult-to-reach areas. Bottle holder uses a universal quick-tightening strap to hold bottles up to 88 mm diameter securely during sampling. Base supports bottom of bottle and provides an adjustable angle.
This item is not returnable. View return policy
Catalog No. NC2171938 Supplier Cole-Parmer Supplier No. 0604205
This modular sampling system allows you to build a sampler according to your application. Select a telescoping aluminum rod (not included) with the length you need, then choose one or more beakers and/or a bottle holder to attach to the rod. Angled and pendulum beakers are available in plastic or metal, and the bottle holder fits plastic, glass, or metal bottles. Angled beakers maintain position while sampling. The polypropylene beakers feature an adjustable angle up to 90°, and the 304 SS beaker has a fixed angle with a lip—perfect for scraping caked solids, sludge, and mud. Pendulum beakers automatically swings into a horizontal position, preventing spills. This type of beaker is ideal for deep, narrow shafts and other difficult-to-reach areas. Bottle holder uses a universal quick-tightening strap to hold bottles up to 88 mm diameter securely during sampling. Base supports bottom of bottle and provides an adjustable angle.