Learn More
Hach Company Bottle Kit-24x 1 Liter PE-AWRS
Supplier:  Hach Company AW241000
Encompass
Bottle Kit-24x 1 Liter PE-for All Weather Refrigerated SamplerAWRS Sampler Bottle Kit with (24) 1 L polyethylene bottles with caps retainer tray and distributor arm
Catalog No. 50-002-08083