Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Chemglass Media Bottle, Square PETG, 1,000 mL, Sterile (CGN-1150-1000)
SDP

Catalog No. 502733510
Encompass_Preferred

Sterile 1,000 mL PETG square media bottle designed for preparation, storage, and transport of laboratory media and reagents. E-beam sterilized to SAL 10^-6 and manufactured to be DNase-, RNase-, and pyrogen-free. Suitable for refrigerated and frozen storage down to -80°C and compatible with routine laboratory handling.

  • Capacity: 1,000 mL.
  • Material: PETG (polyethylene terephthalate glycol-modified).
  • Body style: Square with rounded edges for easier grip.
  • Sterilization: E-beam sterilized to SAL 10^-6.
  • Cleanliness: DNase-free, RNase-free, pyrogen-free.
  • Temperature range: -80°C to 50°C.
  • Closure/thread: 38-430 closure.
  • Packaging: 4 per pack, 12 per case.

Catalog No. 50-273-3510 Supplier Chemglass Life Sciences Supplier No. CGN11501000
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile 1,000 mL PETG square media bottle designed for preparation, storage, and transport of laboratory media and reagents. E-beam sterilized to SAL 10^-6 and manufactured to be DNase-, RNase-, and pyrogen-free. Suitable for refrigerated and frozen storage down to -80°C and compatible with routine laboratory handling.

  • Capacity: 1,000 mL.
  • Material: PETG (polyethylene terephthalate glycol-modified).
  • Body style: Square with rounded edges for easier grip.
  • Sterilization: E-beam sterilized to SAL 10^-6.
  • Cleanliness: DNase-free, RNase-free, pyrogen-free.
  • Temperature range: -80°C to 50°C.
  • Closure/thread: 38-430 closure.
  • Packaging: 4 per pack, 12 per case.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.