Learn More
Chemglass Life Sciences Square Bottle, Media, 1000mL, Sterile, PETG, Graduated, 38-430
SDP
Catalog No. 501532077
Encompass_Preferred
Square Bottle, Media, 1000mL, Sterile, PETG, Graduated, 38-430. Break-resistant and durable Media bottles are manufactured from polyethylene terephthalate copolyester (PETG) and graduated to contain. The bottles are square/octagonal with rounded edges to insure an Easy-Grip for pouring. Cap is manufactured with polyethylene (HDPE) and comes with a security ring which provides a tamper-evident seal. The cap is liner-free and comes with a leak-proof guarantee. Bottles are inexpensive alternatives to glass media bottles. Bottles provide reduced permeability to both CO2 and O2. Bottles are radiation sterilized and non-pyrogenic.
Catalog No. 50-153-2077 Supplier Chemglass Life Sciences Supplier No. CLS11501000
Square Bottle, Media, 1000mL, Sterile, PETG, Graduated, 38-430. Break-resistant and durable Media bottles are manufactured from polyethylene terephthalate copolyester (PETG) and graduated to contain. The bottles are square/octagonal with rounded edges to insure an Easy-Grip for pouring. Cap is manufactured with polyethylene (HDPE) and comes with a security ring which provides a tamper-evident seal. The cap is liner-free and comes with a leak-proof guarantee. Bottles are inexpensive alternatives to glass media bottles. Bottles provide reduced permeability to both CO2 and O2. Bottles are radiation sterilized and non-pyrogenic.