Learn More
Chemglass Life Sciences Bottle, Media, 10000mL, Duran, Clear, with Hose Barb for 1/2" Tubing and GL-45 Vented Cap
SDP
Catalog No. 501294575
Encompass_Preferred
For easy gravity transfer of solutions.¿Duran bottle with drip-free pouring ring and liner-less, one piece, autoclavable, reusable, vented cap.Hose barb outlet at the bottom allows for easy gravity transfer of solutions.The hose barb accepts the ID tubing listed.Other size hose barbs available,.
Catalog No. 50-129-4575 Supplier Chemglass Life Sciences Supplier No. CLS142310L
For easy gravity transfer of solutions.¿Duran bottle with drip-free pouring ring and liner-less, one piece, autoclavable, reusable, vented cap.Hose barb outlet at the bottom allows for easy gravity transfer of solutions.The hose barb accepts the ID tubing listed.Other size hose barbs available,.