Learn More
Chemglass Life Sciences Chemglass Media Bottle, Square PETG, Sterile, 125 mL (CGN-1150-125)
SDP
Catalog No. 502733509
Encompass_Preferred
Sterile 125 mL square PETG media bottle designed for preparing, storing, and transporting cell culture media, reagents, and sterile solutions. The bottle is optically clear, chemically resistant, and suitable for use across a broad laboratory temperature range.
  • Sterile, E-beam sterilized, SAL=10-6.
  • DNase free, RNase free, and pyrogen free.
  • Material PETG (glycol-modified polyethylene terephthalate).
  • Capacity 125 mL.
  • Square shape conserves storage space.
  • Packaged 6 per pack, 48 per case.
  • Temperature range -80°C to 50°C.
Catalog No. 50-273-3509 Supplier Chemglass Life Sciences Supplier No. CGN1150125
Sterile 125 mL square PETG media bottle designed for preparing, storing, and transporting cell culture media, reagents, and sterile solutions. The bottle is optically clear, chemically resistant, and suitable for use across a broad laboratory temperature range.
  • Sterile, E-beam sterilized, SAL=10-6.
  • DNase free, RNase free, and pyrogen free.
  • Material PETG (glycol-modified polyethylene terephthalate).
  • Capacity 125 mL.
  • Square shape conserves storage space.
  • Packaged 6 per pack, 48 per case.
  • Temperature range -80°C to 50°C.