Learn More
Chemglass Life Sciences Chemglass 250 mL PETG Square Media Bottle, Sterile, Nested, Pack of 6
SDP
Catalog No. 502733508
Encompass_Preferred
This 250 mL PETG square media bottle is designed for sterile storage and handling of culture media and critical reagents. It is supplied nested and sterilized to a validated sterility assurance level for single-use laboratory and production workflows.
  • Sterilized by e-beam; SAL = 10^-6.
  • DNase, RNase, and pyrogen free.
  • Temperature range -80°C to 50°C.
  • Packaged nested; 6 per pack, 48 per case.
  • Suitable for storage and handling of media, reagents, and buffers.
Catalog No. 50-273-3508 Supplier Chemglass Life Sciences Supplier No. CGN1150250
This 250 mL PETG square media bottle is designed for sterile storage and handling of culture media and critical reagents. It is supplied nested and sterilized to a validated sterility assurance level for single-use laboratory and production workflows.
  • Sterilized by e-beam; SAL = 10^-6.
  • DNase, RNase, and pyrogen free.
  • Temperature range -80°C to 50°C.
  • Packaged nested; 6 per pack, 48 per case.
  • Suitable for storage and handling of media, reagents, and buffers.