Learn More
Chemglass Life Sciences Chemglass Bottle, Media, 250 mL, Round, GL-45 Screw Cap (CG-8088-250)
SDP
Catalog No. NC2019517
Encompass_Preferred
The 250 mL media storage bottle is a round, clear borosilicate glass vessel designed for laboratory media preparation, storage, and controlled dispensing. It is autoclavable and suitable for storage, mixing, and sampling in laboratory environments.
  • 250 mL capacity
  • Clear borosilicate glass for thermal shock resistance
  • GL-45 screw thread for standard caps
  • Orange polypropylene liner-less screw cap included, autoclavable to 140°C
  • Enhanced white enamel graduations and marking spots
  • Includes a replaceable drip-free polypropylene pour ring
  • Suitable for storage, mixing, and sampling
Catalog No. NC2019517 Supplier Chemglass Life Sciences Supplier No. CG8088250
The 250 mL media storage bottle is a round, clear borosilicate glass vessel designed for laboratory media preparation, storage, and controlled dispensing. It is autoclavable and suitable for storage, mixing, and sampling in laboratory environments.
  • 250 mL capacity
  • Clear borosilicate glass for thermal shock resistance
  • GL-45 screw thread for standard caps
  • Orange polypropylene liner-less screw cap included, autoclavable to 140°C
  • Enhanced white enamel graduations and marking spots
  • Includes a replaceable drip-free polypropylene pour ring
  • Suitable for storage, mixing, and sampling