Learn More
Chemglass Life Sciences Chemglass media bottle, square, 250 mL, sterile PET with HDPE cap, 6/pack
SDP
Catalog No. 502733505
Encompass_Preferred
This 250 mL square media bottle from Chemglass is a sterile, break-resistant PET container designed for preparing, storing, and sampling culture media, buffers, and solutions.
  • Sterile and free of DNase, RNase, and pyrogens.
  • Break-resistant PET construction.
  • High-density polyethylene (HDPE) cap.
  • Square, nestable shape for efficient storage.
  • Packaged 6 per pack, 48 per case.
  • Suitable for media preparation, storage, and sampling.
Catalog No. 50-273-3505 Supplier Chemglass Life Sciences Supplier No. CGN1151250
This 250 mL square media bottle from Chemglass is a sterile, break-resistant PET container designed for preparing, storing, and sampling culture media, buffers, and solutions.
  • Sterile and free of DNase, RNase, and pyrogens.
  • Break-resistant PET construction.
  • High-density polyethylene (HDPE) cap.
  • Square, nestable shape for efficient storage.
  • Packaged 6 per pack, 48 per case.
  • Suitable for media preparation, storage, and sampling.