Learn More
Chemglass Life Sciences Square Bottle, Media, 30mL, Sterile, PETG, Graduated, 20-415
SDP
Catalog No. 50143839
Encompass_Preferred
Square Bottle, Media, 30mL, Sterile, PETG, Graduated, 20-415. • Pyrogen, DNase, RNase & Particle-Free• Transparent & Optically Clear• Freezable at -20°C• Disposable/Recyclable• Gamma Irradiated• Sterility Assurance Level (SAL) 10-6• 20% Overhead Space• Inside Neck Diameter: 34.5mm• Optional Vented Cap AvailableBreak-resistant & durable Media bottles are manufactured from polyethylene terephthalate copolyester (PETG) & graduated to contain. The bottles are square/octagonal with rounded edges to insure an Easy-Grip for pouring. Cap is manufactured with polyethylene (HDPE) & comes with a security ring which provides a tamper-evident seal. The cap is liner-free & comes with a leak-proof guarantee. Bottles are inexpensive alternatives to glass media bottles. Bottles provide reduced permeability to both CO2 & O2. Bottles are radiation sterilized & non-pyrogenic.Bottle Design: Patent US D535566STamper-evident Cap Design: Patent US D561587SBOTTLES ARE NOT AUTOCLAVABLE.
Catalog No. 50-143-839 Supplier Chemglass Life Sciences Supplier No. CLS11500030
Square Bottle, Media, 30mL, Sterile, PETG, Graduated, 20-415. • Pyrogen, DNase, RNase & Particle-Free• Transparent & Optically Clear• Freezable at -20°C• Disposable/Recyclable• Gamma Irradiated• Sterility Assurance Level (SAL) 10-6• 20% Overhead Space• Inside Neck Diameter: 34.5mm• Optional Vented Cap AvailableBreak-resistant & durable Media bottles are manufactured from polyethylene terephthalate copolyester (PETG) & graduated to contain. The bottles are square/octagonal with rounded edges to insure an Easy-Grip for pouring. Cap is manufactured with polyethylene (HDPE) & comes with a security ring which provides a tamper-evident seal. The cap is liner-free & comes with a leak-proof guarantee. Bottles are inexpensive alternatives to glass media bottles. Bottles provide reduced permeability to both CO2 & O2. Bottles are radiation sterilized & non-pyrogenic.Bottle Design: Patent US D535566STamper-evident Cap Design: Patent US D561587SBOTTLES ARE NOT AUTOCLAVABLE.