Learn More
Chemglass Life Sciences Chemglass media bottle, square PETG, sterile, 30 mL, 5/Pack
SDP
Catalog No. 502733512
Encompass_Preferred
Sterile 30 mL square PETG media bottle designed for handling and storing laboratory media and critical reagents. The bottle is e-beam sterilized to a SAL of 10^-6, free of DNase, RNase, and pyrogens, and is freezable from -80°C to 50°C.
  • Sterile, e-beam sterilized to SAL 10^-6.
  • DNase, RNase, and pyrogen free.
  • Made from PETG for transparency and chemical compatibility.
  • 30 mL capacity with square shape for stable, space-efficient storage.
  • Freezable from -80°C to 50°C.
  • Packaged 5 per pack, 40 per case.
Catalog No. 50-273-3512 Supplier Chemglass Life Sciences Supplier No. CGN1150030
Sterile 30 mL square PETG media bottle designed for handling and storing laboratory media and critical reagents. The bottle is e-beam sterilized to a SAL of 10^-6, free of DNase, RNase, and pyrogens, and is freezable from -80°C to 50°C.
  • Sterile, e-beam sterilized to SAL 10^-6.
  • DNase, RNase, and pyrogen free.
  • Made from PETG for transparency and chemical compatibility.
  • 30 mL capacity with square shape for stable, space-efficient storage.
  • Freezable from -80°C to 50°C.
  • Packaged 5 per pack, 40 per case.