Learn More
Chemglass Life Sciences Chemglass Media Bottle, Square PETG, Sterile, 500 mL (CGN-1150-500)
SDP
Catalog No. 502733507
Encompass_Preferred
This 500 mL sterile PETG square media bottle is designed for preparation, storage, and transport of laboratory media, reagents, and biological buffers. It is sterilized to a sterile assurance level (SAL) of 10^-6 and is free of DNase, RNase, and pyrogens, making it suitable for cell culture workflows.
  • 500 mL capacity
  • Made from PETG for optical clarity and durability
  • Square, rounded-edge shape for easy grip and efficient storage
  • E-beam sterilized to a SAL of 10^-6
  • DNase, RNase, and pyrogen free
  • Usable from -80°C to 50°C
  • Packaged: 8 per pack, 24 per case
Catalog No. 50-273-3507 Supplier Chemglass Life Sciences Supplier No. CGN1150500
This 500 mL sterile PETG square media bottle is designed for preparation, storage, and transport of laboratory media, reagents, and biological buffers. It is sterilized to a sterile assurance level (SAL) of 10^-6 and is free of DNase, RNase, and pyrogens, making it suitable for cell culture workflows.
  • 500 mL capacity
  • Made from PETG for optical clarity and durability
  • Square, rounded-edge shape for easy grip and efficient storage
  • E-beam sterilized to a SAL of 10^-6
  • DNase, RNase, and pyrogen free
  • Usable from -80°C to 50°C
  • Packaged: 8 per pack, 24 per case