Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Media Bottle, Square PETG, Sterile, 60 mL (CGN-1150-060)
SDP

Catalog No. 502733511
Encompass_Preferred

This 60 mL square media bottle is made from PETG and supplied sterile for aseptic cell culture, media preparation, reagent storage, and sample handling. It is packaged in nest packs for sterile handling and laboratory convenience.

  • Capacity: 60 mL.
  • Material: PETG.
  • Sterile: sterilized by e-beam; SAL = 10^-6.
  • Free of DNase, RNase, and pyrogens.
  • Packaged in nests; 6 per pack, 48 per case.
  • Temperature range: -80°C to 50°C.
  • Typical use: media and reagent storage and sample handling.

Catalog No. 50-273-3511 Supplier Chemglass Life Sciences Supplier No. CGN1150060
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 60 mL square media bottle is made from PETG and supplied sterile for aseptic cell culture, media preparation, reagent storage, and sample handling. It is packaged in nest packs for sterile handling and laboratory convenience.

  • Capacity: 60 mL.
  • Material: PETG.
  • Sterile: sterilized by e-beam; SAL = 10^-6.
  • Free of DNase, RNase, and pyrogens.
  • Packaged in nests; 6 per pack, 48 per case.
  • Temperature range: -80°C to 50°C.
  • Typical use: media and reagent storage and sample handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.