Learn More
Cole-Parmer Corning Pyrex 1396 Glass Media Bottle, Square, 250 mL; 10/CS
Catalog No. NC1917721 Shop All Cole-Parmer Products
Encompass_Preferred
These space-saving bottles are made of borosilicate glass for good chemical and thermal resistance. Bottles include matching polypropylene caps.
Catalog No. NC1917721 Supplier Cole-Parmer Supplier No. 3459424
These space-saving bottles are made of borosilicate glass for good chemical and thermal resistance. Bottles include matching polypropylene caps.