Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer BOTTLE MEDIA PYREX 2L 10/CS

Catalog No. NC1907623 Shop All Cole-Parmer Products
Encompass_Preferred

Pyrex 1395-2L Media Storage Bottle w/ Screw Cap, 2 L; 10/Pk

Catalog No. NC1907623 Supplier Cole-Parmer Supplier No. 3451426
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyrex 1395-2L Media Storage Bottle w/ Screw Cap, 2 L; 10/Pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.