Learn More
Chemglass Life Sciences COMPLETE 250mL Media Bottle w/Siphon Assembly
SDP
Catalog No. 50143213
Encompass
Standard media bottle with siphon assembly consisting of stainless steel siphon pipe and silicone tubing cut to length.All stainless parts are manufactured from 316L stainless steel electropolished to a 20Ra or better finish.The silicone tubing is peroxide cured and meets 21 CFR 177.2600 material specifications.Assembled, the media bottle with siphon assembly allows the user to create precisely the desired dip tube angle to retrieve virtually all of the contents of fluid from the bottle.
Catalog No. 50-143-213 Supplier Chemglass Life Sciences Supplier No. CLS1420250
Standard media bottle with siphon assembly consisting of stainless steel siphon pipe and silicone tubing cut to length.All stainless parts are manufactured from 316L stainless steel electropolished to a 20Ra or better finish.The silicone tubing is peroxide cured and meets 21 CFR 177.2600 material specifications.Assembled, the media bottle with siphon assembly allows the user to create precisely the desired dip tube angle to retrieve virtually all of the contents of fluid from the bottle.