Learn More
Wilmad Labglass BOTTLE ONLY GAS WASH 500ML
Catalog No. 50807150
Encompass_Preferred
Bottle, Gas Washing, Tall Form, Fritted. Bottle Only. Capacity: 500mL, Joint Size: [ST] 29/42; Porosity: Extra Coarse. Manufactured with 12mm OD x 25mm length fritted cylinder. Supplied with LG-1004-110 plastic joint clamp.
Catalog No. 50-807-150 Supplier Wilmad Labglass Supplier No. LG3762114
Bottle, Gas Washing, Tall Form, Fritted. Bottle Only. Capacity: 500mL, Joint Size: [ST] 29/42; Porosity: Extra Coarse. Manufactured with 12mm OD x 25mm length fritted cylinder. Supplied with LG-1004-110 plastic joint clamp.