Learn More
Foxx Life Sciences EZLabpure Round Wide Mouth Media Bottle with Graduation Marks, PETG, 500mL, GL45 Closed VersaCap, Sterile, 24/CS
GREENER_CHOICE
Catalog No. 502236479
Encompass_Preferred
EZLabpure™ sterile round media bottles (PS and PETG) are designed for Life Science researchers to store solutions such as tissue culture media, serum, and buffers. Their low-profile design improves media flow for smooth, easy pouring, while the narrow shoulder and molded finger grips ensure a secure hold—even with gloved hands.
Each bottle features a leak-proof, full-thread polypropylene GL45 VersaCap for enhanced safety, easy handling, and convenient labeling. The innovative cap design also allows effortless opening and tightening.
PS bottles are optically clear, highly impact-resistant, and capable of withstanding full vacuum. They are compatible with the Autofil2™ filtration system.
Catalog No. 50-223-6479 Supplier Foxx Life Sciences Supplier No. 11772FLS
EZLabpure™ sterile round media bottles (PS and PETG) are designed for Life Science researchers to store solutions such as tissue culture media, serum, and buffers. Their low-profile design improves media flow for smooth, easy pouring, while the narrow shoulder and molded finger grips ensure a secure hold—even with gloved hands.
Each bottle features a leak-proof, full-thread polypropylene GL45 VersaCap for enhanced safety, easy handling, and convenient labeling. The innovative cap design also allows effortless opening and tightening.
PS bottles are optically clear, highly impact-resistant, and capable of withstanding full vacuum. They are compatible with the Autofil2™ filtration system.