Learn More
Med Vet International Amber Plastic Graduated Ovals, 4oz Liquid Bottle, Twist Off Child Resistant Caps, 25/Pack
Catalog No. 503043687
Encompass_Preferred
Amber Plastic Graduated Ovals. Each case of vials is sold with twist off child resistant caps to prevent children from getting into them. Lightweight transparent amber ovals graduated in ounces and milliliters. Colored prescription bottles are ideal for packaging and dispensing light-sensitive liquid medication. This is because of their ability to prevent ultra-violet light from damaging or degrading any photosensitive medication, and they let enough light in to see the contents. Sold in eaches.
Catalog No. 50-304-3687 Supplier Med Vet International Supplier No. 25PKOVAL4
Amber Plastic Graduated Ovals. Each case of vials is sold with twist off child resistant caps to prevent children from getting into them. Lightweight transparent amber ovals graduated in ounces and milliliters. Colored prescription bottles are ideal for packaging and dispensing light-sensitive liquid medication. This is because of their ability to prevent ultra-violet light from damaging or degrading any photosensitive medication, and they let enough light in to see the contents. Sold in eaches.