Learn More
Beckman Coulter Beckman Coulter BOTTLE, PP 29 X 104 50ML (PKG-25)
Catalog No. 501631975
Encompass_Preferred
50 mL Polypropylene Bottle. Dimensions: 29 X 104mm, Package of 25
Catalog No. 50-163-1975 Supplier Beckman Coulter Supplier No. 357003
50 mL Polypropylene Bottle. Dimensions: 29 X 104mm, Package of 25