Learn More
Research Products International Corp Bottle Reagent Dispenser, 5-60ml
SDP
Catalog No. 501977141
Encompass
Precision quality bottle reagent dispenser attaches easily to all common reagent bottles with adapters supplied. Dispenses most reagents (except HF) with an accuracy of ±:0.7% and reproducibility of <0.1%. Constructed of chemical and heat resistant materials for trouble free performance and durability. Entire dispenser can be autoclaved at 121°C without disassembly. Positive locking selector allows precise dispensing over the entire volume range.
Supplied complete with five bottle top adapters (28mm, 32mm, 36mm, 40mm and 45mm diameter) discharge outlet and suction tube. Fits 100ml, 250ml, 500ml, 1000ml, 2500ml and 4 liter bottles.
Catalog No. 50-197-7141 Supplier Research Products International Corp Supplier No. 050501
Precision quality bottle reagent dispenser attaches easily to all common reagent bottles with adapters supplied. Dispenses most reagents (except HF) with an accuracy of ±:0.7% and reproducibility of <0.1%. Constructed of chemical and heat resistant materials for trouble free performance and durability. Entire dispenser can be autoclaved at 121°C without disassembly. Positive locking selector allows precise dispensing over the entire volume range.
Supplied complete with five bottle top adapters (28mm, 32mm, 36mm, 40mm and 45mm diameter) discharge outlet and suction tube. Fits 100ml, 250ml, 500ml, 1000ml, 2500ml and 4 liter bottles.