Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Bottle Rocket Launcher
GREENER_CHOICESCI_ED

Catalog No. S139432 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139432 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139432 Supplier Supertek™ Supplier No. PH10805
Add to Cart
Add to Cart
This item is not returnable. View return policy

Build rockets from ordinary plastic soda bottles that can soar to heights in excess of 150 feet when using the Supertek™ Bottle Rocket Launcher. All you need is water!

  • Comes with 1 Solid aluminum bottle rocket launcher.
  • A quick tug on the release cord and your rocket can soar 150+ feet!
  • Instruction sheet included.

Specifications

Grade 6+
Format Kit
Languages English
Subject Physical Science - Pressure and Volume
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.