Learn More
Cole-Parmer BOTTLE,ROUND,500ML, 24/CS
Supplier:  Cole-Parmer 3521044
Encompass
Environmental Express Nalgene® Wide-Mouth Level 1 Certified Natural Color Bottle, Round, 4 mL Nitric Acid Preserved, 500 mL; 24/CS
Catalog No. NC2161766