Learn More
Chemglass Life Sciences 50mL Bottle, 33-400 GPI Thread, Amber Glass
SDP
Catalog No. CG82001
Encompass_Preferred
50mL Bottle, 33-400 GPI Thread, Amber Glass. These round, wide mouth amber glass bottles are used for storage of light sensitive chemicals. The 50mL bottles have a black phenolic cap with a polyethylene cone shaped liner. Sold with cap attached to the bottle in case quantities only.
Catalog No. CG82001 Supplier Chemglass Life Sciences Supplier No. CG82001
50mL Bottle, 33-400 GPI Thread, Amber Glass. These round, wide mouth amber glass bottles are used for storage of light sensitive chemicals. The 50mL bottles have a black phenolic cap with a polyethylene cone shaped liner. Sold with cap attached to the bottle in case quantities only.