Learn More
Med Vet International Boston Round Yorker Dropper Bottle, 120mL/4 oz., 25/bag
Catalog No. 502643585
Encompass_Preferred
Removable dropper tips allow refilling. Bottle and tips are made of low density polythelene. Screwcap made of polystyrene. 25 per bag.
Catalog No. 50-264-3585 Supplier Med Vet International Supplier No. BSRD120
Removable dropper tips allow refilling. Bottle and tips are made of low density polythelene. Screwcap made of polystyrene. 25 per bag.