Learn More
Chemglass Life Sciences Roux Bottle, 1, 200mL
SDP
Catalog No. 50143682
Encompass_Preferred
Roux Bottle, 1,200mL. The 1,200mL Roux bottle is designed so that when laid down flat, it will hold approximately one-half of the total volume. The bottle is manufactured from Type 1, Class A borosilicate glass. The bottle is useful for growing mass cultures and the offset neck prevents cultures from entering the neck area. Supplied complete with a 38-415 rubber lined closure. Sold 1 per case.Bottle Dimensions: 290mm L x 122mm W x 56mm H
Catalog No. 50-143-682 Supplier Chemglass Life Sciences Supplier No. CLS11951200
Roux Bottle, 1,200mL. The 1,200mL Roux bottle is designed so that when laid down flat, it will hold approximately one-half of the total volume. The bottle is manufactured from Type 1, Class A borosilicate glass. The bottle is useful for growing mass cultures and the offset neck prevents cultures from entering the neck area. Supplied complete with a 38-415 rubber lined closure. Sold 1 per case.Bottle Dimensions: 290mm L x 122mm W x 56mm H