Learn More
Hach Company pH Electrode Storage Bottle
Catalog No. 501997284 Shop All Hach Company Products
Encompass
Store your pH electrodes in this 0.5 oz bottle between uses. Rubber grommet in lid ensures tight seal with 12-mm electrodes.
Catalog No. 50-199-7284 Supplier Hach Company Supplier No. 9260300
Store your pH electrodes in this 0.5 oz bottle between uses. Rubber grommet in lid ensures tight seal with 12-mm electrodes.