Learn More
Hach Company Soaker Bottle for Electrode Storage
Catalog No. 501997148 Shop All Hach Company Products
Encompass
Soaker Bottle for electrode short-term and long-term storage. Using soaker bottle for electrode storage prevents reference junction and/or sensing element from drying out to ensure best performances.
Catalog No. 50-199-7148 Supplier Hach Company Supplier No. 5192900
Soaker Bottle for electrode short-term and long-term storage. Using soaker bottle for electrode storage prevents reference junction and/or sensing element from drying out to ensure best performances.